SY3H3D677B54 Synthonix Corporation

6-Bromoquinazoline-2,4(1H,3H)-dione

  • MDL number MFCD00462868
  • Cas Number : 88145-89-5
  • Molecular Weight : 241.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White to Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47570.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47570.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart