SY3H3D677B57 Synthonix Corporation

3-Bromo-1H-indazole-7-carboxylic acid

  • MDL number MFCD11977526
  • Cas Number : 1040101-01-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Off-white or gray to light yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47573.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47573.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart