SY3H3D677B5D Synthonix Corporation

6-Bromoquinazolin-4-ol

  • MDL number MFCD00115131
  • Cas Number : 32084-59-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47604.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/47604.html
pictograms GHS07
hstatements H302, H315, H319, H332, H335
Availability details for
Quantity
  • Availability
  • Add to Cart