SY3H3D677B69 Synthonix Corporation

5-(4-Bromo-2-fluorophenyl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD09999212
  • Cas Number : 299937-74-9
  • Molecular Weight : 274.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/47632.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47632.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart