SY3H3D677B6E Synthonix Corporation

2-(Bromomethyl)-5-chlorobenzonitrile

  • MDL number No MDL found
  • Cas Number : 50712-69-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/47664.html

Safety Information

MSDS https://synthonix.com/47664.html
Availability details for
Quantity
  • Availability
  • Add to Cart