SY3H3D677B78 Synthonix Corporation

2-Bromo-4-fluoro-5-methylbenzonitrile

  • MDL number MFCD07782070
  • Cas Number : 916792-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47675.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47675.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart