SY3H3D677B82 Synthonix Corporation

2-(3-Bromo-4-fluorophenyl)acetonitrile

  • MDL number No MDL found
  • Cas Number : 501420-63-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/47685.html

Safety Information

MSDS https://synthonix.com/47685.html
Availability details for
Quantity
  • Availability
  • Add to Cart