SY3H3D677BCC Synthonix Corporation

3-Bromobenzofuran

  • MDL number MFCD01029425
  • Cas Number : 59214-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Freezer
Physical Form Pale-yellow to Yellow-brown to Brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47829.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47829.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart