SY3H3D677BD6 Synthonix Corporation

4-bromo-1-methyl-2-(trifluoromethyl)benzene

  • MDL number MFCD01631587
  • Cas Number : 86845-27-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Colorless to Pale-yellow to Yellow-brown Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47847.html

Safety Information

signalword Warning
pcodes P210, P261, P280, P405
MSDS https://synthonix.com/47847.html
pictograms GHS07
hstatements H227, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart