SY3H3D677BE7 Synthonix Corporation

2-(4-Bromophenyl)-2H-1,2,3-triazole

  • MDL number MFCD00603190
  • Cas Number : 74733-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47918.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47918.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart