SY3H3D677BF8 Synthonix Corporation

5-Bromo-2-(methylthio)benzo[d]thiazole

  • MDL number MFCD01935107
  • Cas Number : 203395-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Light-brown to brown powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47955.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47955.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart