SY3H3D677C39 Synthonix Corporation

3-Bromo-4-(hydroxymethyl)benzonitrile

  • MDL number No MDL found
  • Cas Number : 90110-98-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/48079.html

Safety Information

MSDS https://synthonix.com/48079.html
Availability details for
Quantity
  • Availability
  • Add to Cart