SY3H3D677C3F Synthonix Corporation

2-Bromo-4-chlorophenylacetic acid

  • MDL number MFCD09032951
  • Cas Number : 52864-56-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to light-yellow to orange powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48126.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48126.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart