SY3H3D677C5F Synthonix Corporation

1-(4-Bromo-2-chlorophenyl)ethanone

  • MDL number No MDL found
  • Cas Number : 252561-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/48244.html

Safety Information

MSDS https://synthonix.com/48244.html
Availability details for
Quantity
  • Availability
  • Add to Cart