SY3H3D677C67 Synthonix Corporation

1-(2-Bromo-6-fluorophenyl)ethanone

  • MDL number MFCD13193629
  • Cas Number : 928715-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Colorless to Pale-yellow to Yellow-brown Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48252.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48252.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart