SY3H3D677C9D Synthonix Corporation

1-Bromo-3-methoxy-5-(trifluoromethyl)benzene

  • MDL number MFCD09999639
  • Cas Number : 627527-23-5
  • Molecular Weight : 255.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Colorless to Pale-yellow to Yellow-brown Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48265.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48265.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart