SY3H3D677CA8 Synthonix Corporation

4-Bromo-1,2-bis(bromomethyl)benzene

  • MDL number No MDL found
  • Cas Number : 69189-19-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/48300.html

Safety Information

MSDS https://synthonix.com/48300.html
Availability details for
Quantity
  • Availability
  • Add to Cart