SY3H3D677CCE Synthonix Corporation

1H-Benzo[d]imidazole-6-carboxamide

  • MDL number MFCD27978434
  • Cas Number : 116568-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48398.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48398.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart