SY3H3D677CD6 Synthonix Corporation

Benzo[d]oxazol-2-ylmethanol

  • MDL number No MDL found
  • Cas Number : 77186-95-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/48459.html

Safety Information

MSDS https://synthonix.com/48459.html
Availability details for
Quantity
  • Availability
  • Add to Cart