SY3H3D677CE2 Synthonix Corporation

2-Bromo-3-methoxybenzaldehyde

  • MDL number MFCD09834663
  • Cas Number : 10401-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48572.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48572.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart