SY3H3D677D22 Synthonix Corporation

2-Bromo-4-hydroxy-5-methoxybenzaldehyde

  • MDL number MFCD01119128
  • Cas Number : 60632-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48649.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48649.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart