SY3H3D677D36 Synthonix Corporation

1-Bromo-2-chloro-4-ethoxy-3-fluorobenzene

  • MDL number MFCD19440929
  • Cas Number : 909122-17-4
  • Molecular Weight : 253.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48720.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48720.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart