SY3H3D677D45 Synthonix Corporation

1-(1-Bromoethyl)-2-fluorobenzene

  • MDL number MFCD11180278
  • Cas Number : 405931-46-6
  • Molecular Weight : 203.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48765.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48765.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart