SY3H3D677D53 Synthonix Corporation

4-Bromoindoline

  • MDL number MFCD07371633
  • Cas Number : 86626-38-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Pale-yellow to Yellow to Brown Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48872.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48872.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart