SY3H3D677D5C Synthonix Corporation

1-(1-Bromoethyl)-4-nitrobenzene

  • MDL number MFCD11634400
  • Cas Number : 19935-81-0
  • Molecular Weight : 230.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48914.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48914.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart