SY3H3D677D5F Synthonix Corporation

1-Bromo-2,4-dimethyl-5-nitrobenzene

  • MDL number MFCD00075824
  • Cas Number : 69383-59-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48922.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/48922.html
pictograms GHS07
hstatements H302, H312, H315, H319, H332, H335
Availability details for
Quantity
  • Availability
  • Add to Cart