SY3H3D677D86 Synthonix Corporation

1-Bromo-2-ethoxy-4-nitrobenzene

  • MDL number MFCD03790890
  • Cas Number : 423165-33-7
  • Molecular Weight : 246.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to yellow to pale-brown powder or crystals
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/48984.html

Safety Information

signalword Warning
pcodes P261,P305,P351,P338
MSDS https://synthonix.com/48984.html
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart