SY3H3D677DA1 Synthonix Corporation

1-(5-Bromo-2-fluorophenyl)ethanol

  • MDL number MFCD12406116
  • Cas Number : 552331-15-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49140.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49140.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart