SY3H3D677DAB Synthonix Corporation

(R)-1-(4-Bromo-2-fluorophenyl)ethanamine hydrochloride

  • MDL number No MDL found
  • Cas Number : 845829-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49223.html

Safety Information

MSDS https://synthonix.com/49223.html
Availability details for
Quantity
  • Availability
  • Add to Cart