SY3H3D677E08 Synthonix Corporation

Bis(oxazol-4-ylmethyl)amine hydrochloride

  • MDL number MFCD28369488
  • Cas Number : 1646152-51-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49629.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49629.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart