SY3H3D677E12 Synthonix Corporation

4-Bromo-1-cyclopentyl-3-(trifluoromethyl)-1H-pyrazole

  • MDL number MFCD24448783
  • Cas Number : 1437794-36-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Colorless to Pale-yellow to Yellow-brown Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49717.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49717.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart