SY3H3D677E18 Synthonix Corporation

4-Bromo-2,3-dihydro-1H-inden-1-amine hydrochloride

  • MDL number MFCD12761521
  • Cas Number : 1251922-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49765.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49765.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart