SY3H3D677E28 Synthonix Corporation

Bicyclo[4.2.0]octa-1,3,5-trien-7-ylmethanol

  • MDL number MFCD21764776
  • Cas Number : 15100-35-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Colorless to light-yellow liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49879.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49879.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart