SY3H3D677E3A Synthonix Corporation

2-Bromo-6-morpholinopyridine

  • MDL number MFCD07772824
  • Cas Number : 332134-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49991.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49991.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart