SY3H3D677E46 Synthonix Corporation

2-Bromo-1-(methoxymethoxy)-4-methylbenzene

  • MDL number MFCD11850106
  • Cas Number : 104750-60-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50126.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50126.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart