SY3H3D677E88 Synthonix Corporation

1-(5-Bromopyrimidin-2-yl)-3-piperidinol

  • MDL number MFCD08460363
  • Cas Number : 914347-67-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50216.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50216.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart