SY3H3D677E94 Synthonix Corporation

1-(3-Bromophenyl)-N,N-dimethylmethanamine

  • MDL number MFCD04117937
  • Cas Number : 4885-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Colorless to light-yellow to yellow liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50233.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50233.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart