SY3H3D677EA5 Synthonix Corporation

N-Boc-2-Amino-4-cyanothiazole

  • MDL number MFCD12964063
  • Cas Number : 1210278-19-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50329.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50329.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart