SY3H3D678571 Synthonix Corporation

7-Bromo-1,2,3,4-tetrahydronaphthalen-2-amine hydrochloride

  • MDL number MFCD11506000
  • Cas Number : 1187928-01-3
  • Molecular Weight : 262.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/52740.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/52740.html
pictograms GHS07
hstatements H302, H315, H319, H332, H335
Availability details for
Quantity
  • Availability
  • Add to Cart