SY3H3D678579 Synthonix Corporation

N-Boc-D-aspartic Acid

  • MDL number No MDL found
  • Cas Number : 62396-48-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/50478.html

Safety Information

MSDS https://synthonix.com/50478.html
Availability details for
Quantity
  • Availability
  • Add to Cart