SY3H3D67857C Synthonix Corporation

tert-Butyl (5-oxopyrrolidin-3-yl)carbamate

  • MDL number MFCD17677321
  • Cas Number : 1245648-84-3
  • Molecular Weight : 200.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50490.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50490.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart