SY3H3D6785AC Synthonix Corporation

N-Boc-2-methyl-1,3-propanediamine

  • MDL number MFCD06804536
  • Cas Number : 480452-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. refrigerator
Physical Form Clear liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50610.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50610.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart