SY3H3D6785BD Synthonix Corporation

2,4-Bis(trifluoromethyl)benzaldehyde

  • MDL number MFCD00083535
  • Cas Number : 59664-42-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Clear liquid / Colour: Colorless - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50660.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50660.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart