SY3H3D6785D5 Synthonix Corporation

7-Bromo-3-chloroquinoline

  • MDL number No MDL found
  • Cas Number : 1246549-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/50715.html

Safety Information

MSDS https://synthonix.com/50715.html
Availability details for
Quantity
  • Availability
  • Add to Cart