SY3H3D6785DA Synthonix Corporation

2-Bromo-6-fluoroquinoline

  • Cas Number : 159870-91-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/50720.html

Safety Information

MSDS https://synthonix.com/50720.html
Availability details for
Quantity
  • Availability
  • Add to Cart