SY3H3D6785FF Synthonix Corporation

2-(4-Bromophenyl)thiazole

  • MDL number MFCD09997917
  • Cas Number : 27149-27-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50849.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50849.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart