SY3H3D678601 Synthonix Corporation

4-Bromo-5-methylindoline-2,3-dione

  • MDL number MFCD00560709
  • Cas Number : 147149-84-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50865.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50865.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart