SY3H3D678653 Synthonix Corporation

6-Bromo-2-methylquinazolin-4(3H)-one

  • MDL number MFCD00548197
  • Cas Number : 5426-59-5
  • Molecular Weight : 239.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Pale-yellow to Yellow to Brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51123.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51123.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart