SY3H3D67865B Synthonix Corporation

2-Bromo-N-(3-(trifluoromethyl)phenyl)acetamide

  • MDL number MFCD00859566
  • Cas Number : 25625-57-4
  • Molecular Weight : 282.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51238.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51238.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart