SY3H3D67865D Synthonix Corporation

7-Bromochroman-4-one

  • MDL number MFCD09744055
  • Cas Number : 18442-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to yellow-brown or purple solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51249.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51249.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart